Learn More
Invitrogen™ SMCHD1 Monoclonal Antibody (CL4265)

Description
Immunogen sequence: DNGRGMTSKQL NNWAVYRLSK FTRQGDFESD HSGYVRPVPV PRSLNSDISY FGVGGKQAVF FVGQSARMIS KPADSQDVHE LVLSKED Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.
Specifications
Specifications
| Antigen | SMCHD1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL4265 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SMCHD1 |
| Gene Accession No. | A6NHR9 |
| Gene Alias | 4931400A14Rik; AW554188; KIAA0650; mKIAA0650; MommeD1; RGD1307234; SMC hinge domain containing 1; SMC hinge domain-containing protein 1; Smchd1; structural maintenance of chromosomes flexible hinge domain containing 1; structural maintenance of chromosomes flexible hinge domain-containing protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.