Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMCHD1 Monoclonal Antibody (CL4270)
GREENER_CHOICE

Catalog No. PIMA524709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524709 Supplier Invitrogen™ Supplier No. MA524709
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: DNGRGMTSKQL NNWAVYRLSK FTRQGDFESD HSGYVRPVPV PRSLNSDISY FGVGGKQAVF FVGQSARMIS KPADSQDVHE LVLSKED Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.

Required for maintenance of X inactivation in females and hypermethylation of CpG islands associated with inactive X. Involved in a pathway that mediates the methylation of a subset of CpG islands slowly and requires the de novo methyltransferase DNMT3B. Required for DUX4 silencing in somatic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCHD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone CL4270
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMCHD1
Gene Accession No. A6NHR9
Gene Alias 4931400A14Rik; AW554188; KIAA0650; mKIAA0650; MommeD1; RGD1307234; SMC hinge domain containing 1; SMC hinge domain-containing protein 1; Smchd1; structural maintenance of chromosomes flexible hinge domain containing 1; structural maintenance of chromosomes flexible hinge domain-containing protein 1
Gene Symbols SMCHD1
Host Species Mouse
Immunogen Recombinant protein fragment of Human SMCHD1 (Product #RP-98295)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23347
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.