Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMCHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596610 Supplier Invitrogen™ Supplier No. PA596610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTDAARRIYD ETQGRQQVLP LDSIYKKTLP DWKRSLPHFR NGKLYFKPIG DPVFARDLLT FPDNVEHCET VFGMLLGDTI ILDNLDAANH YRKEVVKITH CPTLLTRDGD RIRSNGKFGG LQNKAPPMDK LRGMVFGAPV PKQCLILGEQ IDLLQQYRSA VCKLDSVNKD LNSQLEYLRT PDMRKKKQEL DEHEKNLKLI EEKLGMTPIR KCNDSLRHSP KVETTDCPVP PKRMRREATR QNRIITKTDV; Positive Samples: HL-60, SKOV3, Jurkat, U-251MG, HepG2, Rat brain; Cellular Location: Chromosome.

Required for maintenance of X inactivation in females and hypermethylation of CpG islands associated with inactive X. Involved in a pathway that mediates the methylation of a subset of CpG islands slowly and requires the de novo methyltransferase DNMT3B. Required for DUX4 silencing in somatic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCHD1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SMCHD1
Gene Accession No. A6NHR9
Gene Alias 4931400A14Rik; AW554188; KIAA0650; mKIAA0650; MommeD1; RGD1307234; SMC hinge domain containing 1; SMC hinge domain-containing protein 1; Smchd1; structural maintenance of chromosomes flexible hinge domain containing 1; structural maintenance of chromosomes flexible hinge domain-containing protein 1
Gene Symbols SMCHD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1756-2005 of human SMCHD1 (NP_0561102).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23347, 316732
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.