Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMCHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558788
Description
Immunogen sequence: DNGRGMTSKQ LNNWAVYRLS KFTRQGDFES DHSGYVRPVP VPRSLNSDIS YFGVGGKQAV FFVGQSARMI SKPADSQDVH ELVLSKED Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
Required for maintenance of X inactivation in females and hypermethylation of CpG islands associated with inactive X. Involved in a pathway that mediates the methylation of a subset of CpG islands slowly and requires the de novo methyltransferase DNMT3B. Required for DUX4 silencing in somatic cells.
Specifications
SMCHD1 | |
Polyclonal | |
Unconjugated | |
SMCHD1 | |
4931400A14Rik; AW554188; KIAA0650; mKIAA0650; MommeD1; RGD1307234; SMC hinge domain containing 1; SMC hinge domain-containing protein 1; Smchd1; structural maintenance of chromosomes flexible hinge domain containing 1; structural maintenance of chromosomes flexible hinge domain-containing protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23347 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NHR9 | |
SMCHD1 | |
Recombinant protein corresponding to Human SMCHD1. Recombinant protein control fragment (Product #RP-98295). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction