Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMCO1 Antibody (OTI1H6), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274238C
Description
SMCO1 Monoclonal specifically detects SMCO1 in Human samples. It is validated for Western Blot.Specifications
SMCO1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SMCO1 | |
Full length recombinant protein of human SMCO1 (NP_001071125) produced in E.coli. | |
0.1 mL | |
255798.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1H6 | |
Western Blot | |
C3orf43, Chromosome 3 Open Reading Frame 43, Single-Pass Membrane And Coiled-Coil Domain-Containing Protein 1, Single-Pass Membrane Protein With Coiled-Coil Domains 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction