Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMCO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561594 Supplier Invitrogen™ Supplier No. PA561594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQSDFLYPE NPKRREEVNR LHQQLLDCLS DSFDVTNKLT EVLNMHLGRR LASIEMKRDG TIKENCDLII QAIMK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 79%.

SMCO3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCO3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Smco3
Gene Accession No. A2RU48
Gene Alias C030030A07Rik; C12orf69; C27H12orf69; C5H12orf69; RGD1560652; single pass membrane protein with coiled-coil domains 3; single-pass membrane and coiled-coil domain-containing protein 3; single-pass membrane protein with coiled-coil domains 3; SMCO3
Gene Symbols Smco3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMCO3. Recombinant protein control fragment (Product #RP-97857).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.