Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMCO3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569339
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
SMCO3 is a protein coding gene.
Specifications
SMCO3 | |
Polyclonal | |
Unconjugated | |
Smco3 | |
C030030A07Rik; C12orf69; C27H12orf69; C5H12orf69; RGD1560652; single pass membrane protein with coiled-coil domains 3; single-pass membrane and coiled-coil domain-containing protein 3; single-pass membrane protein with coiled-coil domains 3; SMCO3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
440087 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5-1.0 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A2RU48 | |
Smco3 | |
synthetic peptide directed towards the middle region of human SMCO3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction