Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMCO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559435 Supplier Invitrogen™ Supplier No. PA559435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRQLKGKPKK ETSKDKKERK QAMQEA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

SMCO4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCO4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMCO4
Gene Accession No. Q9NRQ5
Gene Alias C11orf75; C29H11orf75; Fn5; Protein FN5; RGD1309410; single pass membrane protein with coiled-coil domains 4; single-pass membrane and coiled-coil domain-containing protein 4; single-pass membrane protein with coiled-coil domains 4; Smco4; Unknown (protein for MGC:133535); UPF0443 protein C11orf75; UPF0443 protein C11orf75 homolog
Gene Symbols SMCO4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMCO4. Recombinant protein control fragment (Product #RP-97426).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56935
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.