Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMCO4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570929
Description
This target displays homology in the following species: Cow: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%.
SMCO4 is a protein coding gene.
Specifications
SMCO4 | |
Polyclonal | |
Unconjugated | |
SMCO4 | |
C11orf75; C29H11orf75; Fn5; Protein FN5; RGD1309410; single pass membrane protein with coiled-coil domains 4; single-pass membrane and coiled-coil domain-containing protein 4; single-pass membrane protein with coiled-coil domains 4; Smco4; Unknown (protein for MGC:133535); UPF0443 protein C11orf75; UPF0443 protein C11orf75 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
56935 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NRQ5 | |
SMCO4 | |
synthetic peptide directed towards the N-terminal region of human SMCO4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction