Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMCP Antibody (5C10A2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | SMCP |
---|---|
Clone | 5C10A2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Description
SMCP Monoclonal specifically detects SMCP in Human samples. It is validated for Western Blot, ELISA.Specifications
SMCP | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human | |
HSMCSGEN1, MCSMGC26305, MCSPMGC26519, mitochondrial capsule selenoprotein, sperm mitochondria-associated cysteine-rich protein, sperm mitochondrial-associated cysteine-rich protein | |
SMCP | |
IgG2b | |
Affinity Purified |
5C10A2 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Developmental Biology | |
PBS with 0.05% Sodium Azide | |
4184 | |
Purified recombinant fragment of human SMCP (AA: 1-116) expressed in E. Coli. | |
Primary | |
12.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title