Learn More
Invitrogen™ SMCP Monoclonal Antibody (OTI6G9)

Description
SMCP Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SMCP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI6G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SMCP |
| Gene Accession No. | P49901 |
| Gene Alias | HSMCSGEN1; MCS; MCSP; mitochondrial capsule selenoprotein; SMCP; sperm mitochondria associated cysteine rich protein; sperm mitochondria-associated cysteine-rich protein; sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.