Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMCP Mouse anti-Human, Clone: 5C10D8, Abnova™

Catalog No. 89934941 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-941 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-941 Supplier Abnova Corporation Supplier No. MAB12248
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human SMCP.

Sperm mitochondria differ in morphology and subcellular localization from those of somatic cells. They are elongated, flattened, and arranged circumferentially to form a helical coiled sheath in the midpiece of the sperm flagellum. The protein encoded by this gene localizes to the capsule associated with the mitochondrial outer membranes and is thought to function in the organization and stabilization of the helical structure of the sperm's mitochondrial sheath. [provided by RefSeq]

Specifications

Antigen SMCP
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5C10D8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human SMCP.
Dilution ELISA (1:10000) Immunofluorescence (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene Alias HSMCSGEN1/MCS/MCSP/MGC26305/MGC26519
Gene Symbols SMCP
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human SMCP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4184
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.