Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMCP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557156 Supplier Invitrogen™ Supplier No. PA557156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHSKCCPAKG NQCCPPQQNQ CCQSKGNQCC PPKQNQCCQP KGSQCCPPKH NHCCQPKPPC CIQARCCGLE TKPEVSPLNM ESEPNSPQ Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 39%.

Involved in sperm motility. Its absence is associated with genetic background dependent male infertility. Infertility may be due to reduced sperm motility in the female reproductive tract and inability to penetrate the oocyte zona pellucida.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMCP
Gene Accession No. P49901
Gene Alias HSMCSGEN1; MCS; MCSP; mitochondrial capsule selenoprotein; SMCP; sperm mitochondria associated cysteine rich protein; sperm mitochondria-associated cysteine-rich protein; sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161
Gene Symbols SMCP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMCP. Recombinant protein control fragment (Product #RP-97581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.