Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMCR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559950 Supplier Invitrogen™ Supplier No. PA559950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPAALSQPVL PLAPSSSAPE GPAETDPEVT PQLSSPAPLC LTLQERLLAF ERDRVTIPAA QVALAKQLAG DIALELQAYF RSKFPELPFG AFVPGGPLYD GLQAGAADHV RLL Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 69%.

This gene encodes an outer mitochondrial membrane protein that functions in the regulation of mitochondrial morphology. It can directly recruit the fission mediator dynamin-related protein 1 (Drp1) to the mitochondrial surface. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCR7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIEF2
Gene Accession No. Q96C03
Gene Alias AI482195; Gm11; Mid49; MIEF2; mitochondrial dynamic protein MID49; mitochondrial dynamic protein of 49 kDa; mitochondrial dynamic protein of 49 kDa homolog; mitochondrial dynamics protein MID49; Mitochondrial dynamics protein of 49 kDa; mitochondrial dynamics protein of 49 kDa homolog; Mitochondrial elongation factor 2; RGD1560728; SMCR7; Smith-Magenis syndrome chromosomal region candidate gene 7 protein; smith-Magenis syndrome chromosomal region candidate gene 7 protein homolog; Smith-Magenis syndrome chromosome region, candidate 7; Smith-Magenis syndrome chromosome region, candidate 7 homolog
Gene Symbols MIEF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMCR7. Recombinant protein control fragment (Product #RP-90491).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.