Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMCR7L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113393
Description
SMCR7L Polyclonal Antibody for Western Blot, ELISA
Mitochondrial outer membrane protein which regulates mitochondrial fission. Promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface independently of the mitochondrial fission FIS1 and MFF proteins. Regulates DNM1L GTPase activity and DNM1L oligomerization. Binds ADP and can also bind GDP, although with lower affinity. Does not bind CDP, UDP, ATP, AMP or GTP. Inhibits DNM1L GTPase activity in the absence of bound ADP. Requires ADP to stimulate DNM1L GTPase activity and the assembly of DNM1L into long, oligomeric tubules with a spiral pattern, as opposed to the ring-like DNM1L oligomers observed in the absence of bound ADP. Does not require ADP for its function in recruiting DNM1L. [UniProt].
Specifications
SMCR7L | |
Polyclonal | |
Unconjugated | |
MIEF1 | |
A230016E22; AI452372; dJ1104E15.3; HSU79252; MID51; Mief1; mitochondrial dynamic protein MID51; mitochondrial dynamic protein of 51 kDa; mitochondrial dynamic protein of 51 kDa homolog; mitochondrial dynamics protein MID51; Mitochondrial dynamics protein of 51 kDa; mitochondrial dynamics protein of 51 kDa homolog; Mitochondrial elongation factor 1; Smcr7l; SMCR7-like protein; smith-Magenis syndrome chromosomal region candidate gene 7 protein-like; Smith-Magenis syndrome chromosome region, candidate 7-like | |
Rabbit | |
Protein G | |
RUO | |
239555, 54471 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8BGV8, Q9NQG6 | |
MIEF1 | |
Recombinant Human Mitochondrial dynamics protein MID51 protein (51-309AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction