Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMCR8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117431
Description
Centrifuge briefly prior to opening.
Component of the C9orf72-SMCR8 complex, a complex that has guanine nucleotide exchange factor (GEF) activity and regulates autophagy. In the complex, C9orf72 and SMCR8 probably constitute the catalytic subunits that promote the exchange of GDP to GTP, converting inactive GDP-bound RAB8A and RAB39B into their active GTP-bound form, thereby promoting autophagosome maturation. The C9orf72-SMCR8 complex also acts as a negative regulator of autophagy initiation by interacting with the ATG1/ULK1 kinase complex and inhibiting its protein kinase activity. Acts as a regulator of mTORC1 signaling by promoting phosphorylation of mTORC1 substrates. In addition to its activity in the cytoplasm within the C9orf72-SMCR8 complex, SMCR8 also localizes in the nucleus, where it associates with chromatin and negatively regulates expression of suppresses ULK1 and WIPI2 genes.
Specifications
| SMCR8 | |
| Polyclonal | |
| Unconjugated | |
| SMCR8 | |
| 2310076G09Rik; AI642055; D030073L15Rik; FLJ34716; Guanine nucleotide exchange protein SMCR8; RGD1564621; SMCR8; Smith-Magenis syndrome chromosomal region candidate gene 8 protein; Smith-Magenis syndrome chromosomal region candidate gene 8 protein homolog; Smith-Magenis syndrome chromosome region, candidate 8; Smith-Magenis syndrome chromosome region, candidate 8 homolog; Smith-Magenis syndrome chromosome region, candidate 8 homolog (human) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140775, 497918 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.12 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8TEV9 | |
| SMCR8 | |
| Recombinant protein encompassing a sequence within the center region of human SMCR8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction