Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMDT1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA556934
Description
Immunogen sequence: RWLVLAPVRS GALRSGPSLR KDGDVSAAWS GSGRSLVPSR SVIVTRHighest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 57%.
Specifications
SMDT1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4I9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91689 | |
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
SMDT1 | |
SMDT1 | |
Recombinant protein corresponding to Human SMDT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction