Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMEK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551724 Supplier Invitrogen™ Supplier No. PA551724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGEAVVSPSD KTKNDDDIMD PISKFMERKK LKESEEKEVL LKTNLSGRQS PSFKLSLSSG TKTNLTSQSS TTNLPGSPGS PGSPGSPGSP GSVPKNTSQT AAITTKGGLV GLVDYPDDDE DDDEDEDK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

Regulatory subunit of serine/threonine-protein phosphatase 4. May regulate the activity of PPP4C at centrosomal microtubule organizing centers. The PPP4C-PPP4R2-PPP4R3A PP4 complex specifically dephosphorylates H2AFX phosphorylated on 'Ser-140' (gamma-H2AFX) generated during DNA replication and required for DNA DSB repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMEK1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP4R3A
Gene Accession No. Q6IN85
Gene Alias 1110034C04Rik; BC064465; FLFL1; KIAA2010; mKIAA2010; MSTP033; PP4R3; PP4R3A; Ppp4r3a; protein phosphatase 4 regulatory subunit 3A; protein phosphatase 4, regulatory subunit 3A; RGD1309059; serine/threonine-protein phosphatase 4 regulatory subunit 3A; SMEK homolog 1; SMEK homolog 1, suppressor of mek1; SMEK homolog 1, suppressor of mek1 (Dictyostelium); SMEK1; smk1; smk-1
Gene Symbols PPP4R3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMEK1. Recombinant protein control fragment (Product #RP-89035).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55671
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.