Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMEK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518784
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518784 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518784 Supplier Thermo Scientific Supplier No. PA518784
Add to Cart
Add to Cart

SMEK1 Polyclonal Antibody

SMEK1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SMEK1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PPP4R3A
Gene Accession No. Q6IN85
Gene Alias 1110034C04Rik; BC064465; FLFL1; KIAA2010; mKIAA2010; MSTP033; PP4R3; PP4R3A; Ppp4r3a; protein phosphatase 4 regulatory subunit 3A; protein phosphatase 4, regulatory subunit 3A; RGD1309059; serine/threonine-protein phosphatase 4 regulatory subunit 3A; SMEK homolog 1; SMEK homolog 1, suppressor of mek1; SMEK homolog 1, suppressor of mek1 (Dictyostelium); SMEK1; smk1; smk-1
Gene Symbols PPP4R3A
Host Species Goat
Immunogen Synthetic peptide sequence (EQQRERQDNPKLD) corresponding to the internal amino acids of SMEK1 (aa 645-658).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55671
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.