Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMEK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551545 Supplier Invitrogen™ Supplier No. PA551545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQTFKGLKTK YEQEKDRQNQ KLNSVPSILR SNRFRRDAKA LEEDEEMWFN EDEEEEGKAV MPPLE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

Regulatory subunit of serine/threonine-protein phosphatase 4 (PP4). May regulate the activity of PPP4C at centrosomal microtubule organizing centers. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMEK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP4R3B
Gene Accession No. Q5MIZ7, Q922R5
Gene Alias AW011752; AW557776; FLFL2; KIAA1387; mKIAA1387; Pp4r3b; PPP4R3B; protein phosphatase 4 regulatory subunit 3B; protein phosphatase 4, regulatory subunit 3B; PSY2; RGD1309450; Serine/threonine-protein phosphatase 4 regulatory subunit 3B; SMEK homolog 2; SMEK homolog 2, suppressor of mek1; SMEK homolog 2, suppressor of mek1 (Dictyostelium); SMEK2; smk1
Gene Symbols PPP4R3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMEK2. Recombinant protein control fragment (Product #RP-92005).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104570, 360993, 57223
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.