Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMG1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002320DL594
Description
SMG1 Polyclonal antibody specifically detects SMG1 in Human samples. It is validated for Western BlotSpecifications
SMG1 | |
Polyclonal | |
Western Blot | |
ATXserine/threonine-protein kinase SMG1, EC 2.7.11, EC 2.7.11.1, hSMG-1,61E3.4, KIAA0421phosphatidylinositol 3-kinase-related protein kinase, Lambda/iota protein kinase C-interacting protein, Lambda-interacting protein, LIPPI-3-kinase-related kinase SMG-1, phosphatidylinositol 3-kinase-related kinase, SMG-1, SMG1 homolog, phosphatidylinositol 3-kinase-related kinase (C. elegans) | |
The immunogen recognized by this antibody maps to a region between residues 1625 and 1675 of human PI-3-Kinase-related kinase SMG1 using the numbering given in entry NP_055907.3 (GeneID 23049). | |
0.1 mL | |
Protein Kinase | |
23049 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction