Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMG1 Antibody (SR1684), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | SMG1 |
---|---|
Clone | SR1684 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
SMG1 Monoclonal antibody specifically detects SMG1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SMG1 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ATXserine/threonine-protein kinase SMG1, EC 2.7.11, EC 2.7.11.1, hSMG-1,61E3.4, KIAA0421phosphatidylinositol 3-kinase-related protein kinase, Lambda/iota protein kinase C-interacting protein, Lambda-interacting protein, LIPPI-3-kinase-related kinase SMG-1, phosphatidylinositol 3-kinase-related kinase, SMG-1, SMG1 homolog, phosphatidylinositol 3-kinase-related kinase (C. elegans) | |
A synthesized peptide derived from human SMG1 (Uniprot #: Q96Q15) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1684 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
23049 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title