Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581547 Supplier Invitrogen™ Supplier No. PA581547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SMG1.

This gene encodes a protein involved in nonsense-mediated mRNA decay as part of the mRNA surveillance complex. The protein has kinase activity and is thought to function in NMD by phosphorylating the regulator of nonsense transcripts 1 protein. Alternative spliced transcript variants have been described, but their full-length natures have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMG1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SMG1
Gene Accession No. Q96Q15
Gene Alias 2610207I05Rik; 5430435M13Rik; 61E3.4; ATX; C130002K18Rik; hSMG-1; KIAA0421; lambda/iota protein kinase C-interacting protein; Lambda-interacting protein; LIP; mKIAA0421; phosphatidylinositol 3-kinase-related protein kinase; PI-3-kinase-related kinase SMG-1; RGD1563508; serine/threonine-protein kinase SMG1; SMG1; SMG-1; smg-1 homolog, phosphatidylinositol 3-kinase-related kinase; SMG1 homolog, phosphatidylinositol 3-kinase-related kinase (C. elegans); SMG1 phosphatidylinositol 3-kinase-related kinase; SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Gene Symbols SMG1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human SMG1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23049
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.