Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544295 Supplier Invitrogen™ Supplier No. PA544295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSQGPPTGES SEPEAKVLHT KRLYRAVVEA VHRLDLILCN KTAYQEVFKP Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rat: 86%.

SMG5 plays a role in nonsense-mediated mRNA decay. SMG5 does not have RNase activity by itself. SMG5 promotes dephosphorylation of RENT1. SMG5 is necessary for TERT activity.SMG5 is involved in nonsense-mediated mRNA decay (Ohnishi et al., 2003 [PubMed 14636577]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMG5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SMG5
Gene Accession No. Q9UPR3
Gene Alias BC024683; EST1 telomerase component homolog B; Est1b; EST1-like protein B; Est1p-like protein B; ever shorter telomeres 1B; FLJ34864; hSMG-5; Kiaa1089; LPTS interacting protein; LPTS-interacting protein; LPTSRP1; LPTS-RP1; mKIAA1089; Protein SMG5; RP11-54H19.7; Smg5; SMG-5; SMG-5 homolog; SMG5 homolog nonsense mediated mRNA decay factor; Smg-5 homolog, nonsense mediated mRNA decay factor; Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG5 nonsense mediated mRNA decay factor; SMG5, nonsense mediated mRNA decay factor
Gene Symbols SMG5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SMG5 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23381
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.