Learn More
SMG5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SMG5 |
---|---|
Concentration | 133 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8307
![]() ![]() |
Proteintech Group Inc
126941AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The SMG5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SMG5. This antibody recognizes human antigen. The SMG5 antibody has been validated for the following applications: ELISA analysis.
Specifications
SMG5 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG |
133 μg/mL | |
Polyclonal | |
Rabbit | |
23381 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.