Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMG6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560165 Supplier Invitrogen™ Supplier No. PA560165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEGLERVRI SASELRGILA TLAPQAGSRE NMKELKEARP RKDNRRPDLE IYKPGLSRLR NKPKIKEPPG SEEFKDEIVN DRDCSAVENG TQPVKDVCKE LNNQEQNGPI DP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

This gene encodes a component of the telomerase ribonucleoprotein complex responsible for the replication and maintenance of chromosome ends. The encoded protein also plays a role in the nonsense-mediated mRNA decay (NMD) pathway, providing the endonuclease activity near the premature translation termination codon that is needed to initiate NMD. Alternatively spliced transcript variants encoding distinct protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMG6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMG6
Gene Accession No. Q86US8
Gene Alias AI317223; AU041178; C17orf31; EST1 telomerase component homolog A; EST1A; EST1-like protein A; ever shorter telomeres 1A; hSmg5/7a; Kiaa0732; LRRGT00040; RGD1309609; Smg6; SMG-6; Smg-6 homolog; smg-6 homolog, nonsense mediated mRNA decay factor; Smg-6 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG6 nonsense mediated mRNA decay factor; SMG6, nonsense mediated mRNA decay factor; telomerase subunit EST1A; Telomerase-binding protein EST1A
Gene Symbols SMG6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMG6. Recombinant protein control fragment (Product #RP-91833).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23293
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.