Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101223 Supplier Invitrogen™ Supplier No. PA5101223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SMG7.

SMG7 is involved in nonsense-mediated mRNA decay (Fukuhara et al., 2005 [PubMed 15721257]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMG7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SMG7
Gene Accession No. Q5RJH6, Q92540
Gene Alias 9430023P16Rik; breast cancer-associated antigen SGA-56M; C1orf16; EST1 telomerase component homolog C; Est1c; EST1-like protein C; ever shorter telomeres 1C; hSMG-7; KIAA0250; mKIAA0250; Protein SMG7; SGA56M; Smg7; SMG-7 homolog; smg-7 homolog, nonsense mediated mRNA decay factor; Smg-7 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG7 nonsense mediated mRNA decay factor; SMG7, nonsense mediated mRNA decay factor
Gene Symbols SMG7
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMG7(Accession Q92540), corresponding to amino acid residues S528-T578.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226517, 9887
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.