Learn More
SMG7 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302170A

Description
The recommended shelf life for this product is 1 year from date of receipt.
SMG7 is involved in nonsense-mediated mRNA decay (Fukuhara et al., 2005 [PubMed 15721257]).Specifications
SMG7 | |
Polyclonal | |
Unconjugated | |
SMG7 | |
C1orf16, EST1C, SGA56M, EST1 telomerase component homolog C, EST1-like protein C, breast cancer-associated antigen SGA-56M, ever shorter telomeres 1C, protein SMG7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
226517, 9887 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5RJH6, Q92540 | |
SMG7 | |
Between 1087 and 1137 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.