Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMG9 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB254613 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254613 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB254613 Supplier Novus Biologicals Supplier No. NBP126595AF594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMG9 Polyclonal antibody specifically detects SMG9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen SMG9
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias chromosome 19 open reading frame 61, DKFZp564H1322, F17127_1, FLJ12886, protein SMG9, Protein smg-9 homolog, SMG9
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human suppressor with morphogenetic effect on genitalia protein 9 using the numbering given in entry NP_061981.2 (GeneID 56006).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56006
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.