Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMIM13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23807425UL
Description
SMIM13 Polyclonal specifically detects SMIM13 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SMIM13 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
P0DJ93 | |
SMIM13 | |
This antibody was developed against a recombinant protein corresponding to amino acids: WHLFLSKFKFLRELVGDTGSQEGDHEPSGSETEEDTSSSPHRIRSARQRRAPADEGHRPLT | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C6orf228, Chromosome 6 Open Reading Frame 228, Small Integral Membrane Protein 13, UPF0766 Protein C6orf228 | |
Rabbit | |
Affinity Purified | |
RUO | |
221710 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction