Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMIM17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567209 Supplier Invitrogen™ Supplier No. PA567209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQSLRPEQTRG LLEPERTKTL LPRESRAWEK PPHPACTKDW EAVEVGASS Highest antigen sequence identity to the following orthologs - mouse 66%, rat 64%.

SMIM17 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMIM17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMIM17
Gene Accession No. P0DL12
Gene Alias Small integral membrane protein 17; SMIM17
Gene Symbols SMIM17
Host Species Rabbit
Immunogen Recombinant Human SMIM17. Recombinant protein control fragment (Product #RP-107844).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.