Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMIM24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565991 Supplier Invitrogen™ Supplier No. PA565991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTAKEGESNLG LDLEEKEPGD HERAKSTVM Highest antigen sequence identity to the following orthologs - mouse 50%, rat 53%.

SMIM24 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMIM24
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMIM24
Gene Accession No. O75264
Gene Alias C19orf77; HSPC323; Small integral membrane protein 24; SMIM24; transmembrane protein C19orf77; transmembrane protein HSPC323
Gene Symbols SMIM24
Host Species Rabbit
Immunogen Recombinant Human SMIM24. Recombinant protein control fragment (Product #RP-106675).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284422
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.