Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMIM5 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298123C
Description
SMIM5 Polyclonal specifically detects SMIM5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SMIM5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
C17orf109, Chromosome 17 Open Reading Frame 109, PP12104, Small Integral Membrane Protein 5 | |
Produced in rabbits immunized with E. coli-derived Human SMIM5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
643008 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction