Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMIM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566766
Description
Immunogen sequence: MAATDFVQEMR AVGERLLLKL QRLPQAEPVE I Highest antigen sequence identity to the following orthologs - mouse 84%, rat 78%.
SMIM5 is a protein coding gene.
Specifications
SMIM5 | |
Polyclonal | |
Unconjugated | |
SMIM5 | |
C17orf109; PP12104; Small integral membrane protein 5; SMIM5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
643008 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q71RC9 | |
SMIM5 | |
Recombinant Human SMIM5. Recombinant protein control fragment (Product #RP-107423). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction