Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smith Antigen Antibody (Y12), Novus Biologicals™
SDP

Catalog No. NB600546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600546 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600546 Supplier Novus Biologicals Supplier No. NB600546
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

Smith Antigen Monoclonal specifically detects Smith Antigen in Human, Mouse, Rat, Porcine, Drosophila, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunomicroscopy.

Specifications

Antigen Smith Antigen
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunomicroscopy
Classification Monoclonal
Clone Y12
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.0 μg/mL, ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:100, Immunomicroscopy 1:10 - 1:500
Gene Alias Smith antigen
Host Species Mouse
Immunogen The immunogen is generated from human Smith Antigen.
Purification Method Protein G purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Ab-1 recognizes cross-reactive epitopes on the B'/B and D polypeptides of Sm. Ab-1 precipitates the small nuclear RNAs U1, U2, U4, U5, and U6 providing direct evidence that the Sm antigen resides on all these RNA-protein complexes.
Target Species Human, Mouse, Rat, Pig, Drosophila, Rabbit
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.