Description
- Safety eyewear with a sporty sunglasses design
- Contoured temples for a precise and secure fit
- Ribbed nose pad to prevent slippage
- Clear lenses allow maximum visible light transmission; ideal for indoor use
- Hardcoated lenses for scratch resistance and longer life
- Lenses are made of a polycarbonate material that provides 99.9% UVA/UVB/UVC protection
- Meets ANSI Z87.1+ standards for impact
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance | Meets ANSI Z87.1+ standards for impact |
| Lens Tint | Clear |
| Product Type | Safety Glasses |
| Includes | Ribbed nose pad |
| No. per Pack | 12/Pk. |
| Lens Material | Polycarbonate |