Learn More
Ansell Smith & Wesson™ Magnum™ 3G Safety Glasses
Magnum 3G™ Safety Glasses are popular for use as shooting range glasses, gun glasses, gun range glasses or gun safety glasses
Supplier: Ansell 19955
This item is not returnable.
View return policy
Description
- Meet ANSI Z87.1+ standards and provide 99.9% UVA/UVB/UVC protection
- Platinum frames have a self-adjusting nose pad with rubber fingers, which allows air flow and sweat to dissipate
- Polycarbonate lenses that help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Ergonomically dimensioned three-point extendable temples to adjust fit
- Cut-to-shape lens provides less distortion around the edges
Specifications
Anti-fog | |
3-Point fit | |
Safety glasses | |
Black |
Meets ANSI Z87.1+ standards | |
Smoke | |
Eye protection | |
Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers |