Learn More
Ansell Smith & Wesson™ Magnum™ 3G Safety Glasses
Magnum 3G™ Safety Glasses are popular for use as shooting range glasses, gun glasses, gun range glasses or gun safety glasses
$278.31 - $398.31
Specifications
Description | 3-Point fit |
---|---|
For Use With (Application) | Eye protection |
Frame Color | Black |
Catalog Number | Mfr. No. | Frame Color | Includes | Lens Tint | Lens Coating | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Includes | Lens Tint | Lens Coating | Price | Quantity & Availability | ||
19-130-0897
|
Ansell
19826 |
Black | Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers | Yellow | Uncoated |
Case of 12 for $278.31
|
|
||
19-130-0899
|
Ansell
19794 |
Black | Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers | Clear | Anti-fog |
Dozen of 12 for $398.31
|
|
||
19-130-0898
|
Ansell
19955 |
Black | Built-in hinge; Self-adjusting ergonomic rubber nose pad with rubber fingers | Smoke | Anti-fog | N/A | N/A | N/A | |
Description
- These Smith & Wesson Magnum 3G Safety Glasses have a black frame and IRUV Shade 5.0 lenses.
- Lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB)
- Designed for medium cutting and brazing applications
- Meet ANSI Z87.1+ standards
- Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage
- Ergonomically dimensioned three-point extendable temples – so you can adjust to fit
- Self-adjusting nose pad with rubber fingers designed to prevent glasses from slipping down the nose
- Cut-to-shape lens provides less distortion around the edges
Specifications
3-Point fit | |
Eye protection | |
Black |