Learn More
Ansell Smith & Wesson™ Magnum™ 3G Safety Glasses
Catalog No. 19804278 Shop All Ansell Products
Change view
Click to view available options
Orange lens tint
Catalog No.
19-804-278 Orange lens tint
1 options
This item is not returnable. View return policy
Catalog No. 19-804-278 Supplier Ansell Supplier No. 19829
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Magnum™ 3G Safety Glasses are ergonomically designed with a self-adjusting nose pad.
  • Orange lenses have light-gathering properties for low-contrast environments
  • Meet ANSI Z87.1+ standards
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Ergonomically dimensioned three-point extendable temples for an adjustable fit
  • Self-adjusting nose pad with rubber fingers designed to prevent glasses from slipping down the nose
  • Cut-to-shape lens provides less distortion around the edges
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Impact-resistant, UV-treated
Certifications/Compliance Meets ANSI Z87.1+ standards
Description 3-Point fit
Lens Tint Orange
Product Type Safety glasses
For Use With (Application) Gun safety glasses
Frame Color Platinum
Includes Built-in hinge
No. per Case 12/Cs.
Lens Material Polycarbonate
Frame Material Plastic
UV Protection 0.999