Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/23.5IU Kits

Click to view available options
Quantity:
100/Cs.
200/Cs.
Product Type:
Device, 2nd needle, Support kt
FP, NP Device, Support Kit
FP, NP Device, Support Kit pp
FP, PL Device, Support Kit, pp
Filter-Pro, Needle-Pro Device
Filter-Pro, Point-Lok Device
Needle Gauge:
23 to 25 G
25 G
Description
Pro-Vent™ Plus 1 mL Arterial Blood Sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results.The syringes are available with a broad choice of product features.
- Advanced-formula dry lithium heparin
- Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
- Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
- Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
- Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
- Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
- Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
†Polybagged.
Specifications
Specifications
Needle Gauge | 25 G |
For Use With (Application) | Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results |
Includes | 1mL luer-slip syringe with Filter-Pro™ device, 25G x 0.62 in. preattached needle with Needle-Pro™ needle safety device, support kit (Alcohol prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandag |
Product Type | FP, NP Device, Support Kit pp |
Quantity | 100/Cs. |
Packaging | Sterile Tyvek™ |
Volume (Metric) | 1 mL |
Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction