Learn More
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/7IU Kits
Catalog No. 22024944
Encompass
Click to view available options
Quantity:
100/Cs.
200/Cs.
Product Type:
FP, PL Device, support kit, pp
Filter-Pro Device, Polybag
Filter-Pro, Needle Pro Device
Filter-Pro, Point-Lok Device
Packaging:
Polybag
Sterile Tyvek
Pro-Vent™ Plus 1 mL Arterial Blood Sampling syringes utilize dry lithium heparin with a low concentration of 7 I.U. per mL. The low heparin concentration allows for healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte, and metabolite testing with reduced heparin interference and extended clotting time.
  • Advanced-formula dry lithium heparin
  • Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
  • Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
  • Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
  • Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
  • Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
  • Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
†Polybagged.
Specifications
Needle Gauge 25 G
For Use With (Application) Allows healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte and metabolite testing with reduced heparin interference and extended clotting time
Includes 1mL luer-slip syringe, Filter-Pro™ device, 25G x 0.62 in. preattached needle, Point-Lok™ needle safety device, support kit, and iodophor prep pad
Product Type Filter-Pro, Needle Pro Device
Quantity 200/Cs.
Packaging Sterile Tyvek
Volume (Metric) 1 mL
Sterility Sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.