Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 3cc/21IU Kits
Click to view available options
Quantity:
100/Cs.
200/Cs.
Needle Gauge:
22 to 23 G
23 G
Product Type:
FP, NP, 2nd needle, spprt kt
Filter-Pro, Needle-Pro Device
Description
Pro-Vent™ Plus 3 mL Arterial Blood Sampling syringes utilize dry lithium heparin with a low concentration of 7 I.U. per mL. The low heparin concentration allows for healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte, and metabolite testing with reduced heparin interference and extended clotting time.
- Advanced-formula dry lithium heparin
- Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
- Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
- Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
- Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
- Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
- Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
†Polybagged.
Specifications
Specifications
| Needle Gauge | 22 to 23 G |
| For Use With (Application) | Allows healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte and metabolite testing with reduced heparin interference and extended clotting time |
| Includes | 3mL luer-slip syringe with Filter-Pro™ device, 23G x 1 in. preattached needle with Needle-Pro™ needle safety device, 22 x 1.5 in. secondary needle with Needle-Pro™ needle safety device, support kit |
| Product Type | Filter-Pro, Needle-Pro Device |
| Quantity | 100/Cs. |
| Packaging | Sterile Tyvek™ |
| Volume (Metric) | 3 mL |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction