Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 3cc/70.5IU Kits
Click to view available options
Quantity:
100/Cs.
200/Cs.
Product Type:
Arterial Blood Sampling Kit
FP, NP LL, 2nd needle, kit, pp
FP, NP LS device, kit, pp
FP, NP LS device, support kit
FP, NP LS, pad, label, ice bag
FP, PL LL device, kit, pp
FP, PL LL, 2nd needle, kit, pp
FP,NP LS, 2nd needle, kit, pp
Filter-Pro, Needle-Pro LL dev
Filter-Pro, Needle-Pro LS dev
Filter-Pro, Point-Lok LL dev
Needle Gauge:
22 G
22 to 23 G
23 G
Packaging:
Sterile Tyvek
Description
- Advanced-formula dry lithium heparin
- Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
- Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
- Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
- Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
- Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
- Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
†Polybagged.
Specifications
Specifications
| Needle Gauge | 23 G |
| For Use With (Application) | Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results |
| Includes | 3mL luer-slip syringe with Filter-Pro™ device, 22G x 1 in. preattached needle with Needle-Pro™ needle safety device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant |
| Product Type | Filter-Pro, Needle-Pro LS dev |
| Quantity | 200/Cs. |
| Packaging | Sterile Tyvek |
| Volume (Metric) | 3 mL |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction