Learn More
ICU Medical Pulsator™ Plus Liquid Heparin Arterial Blood Sampling Kits
Catalog No. 22024974
Change view
Click to view available options
Quantity:
100/Cs.
Product Type:
Heparin, 20, 22G Needle
Heparin, 22, 23G Needle
Packaging:
Polybag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Packaging
22-024-974 100/Cs. Heparin, 22, 23G Needle Polybag
22-004-198 100/Cs. Heparin, 20, 22G Needle Polybag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-024-974 Supplier ICU Medical Supplier No. 40391
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Includes
3-cc or 5-cc Luer lock syringe and Filter-Pro™ device in polybag or Tyvek packaging.
Quick-filling syringes respond to arterial pressure, aspirate easily
  • Pulsating effect allows for precise positioning in arterial lumen
  • Syringe response to arterial pressure helps ensure arterial sample
  • Rapid fill minimizes patient discomfort
  • 3mL arterial blood-sampling syringes are easily aspirated during low blood pressure or code situation
  • Needles with different gauges and lengths
  • Prefilled with liquid sodium heparin concentration of 31 IU/mL
  • Specially formulated electrolyte-balanced liquid sodium heparin offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels
  • Clot-free blood sampling without risking sample dilution
  • Provides consistent and accurate analysis for all parameters tested
Order Info
  • With or without support kit (alcohol prep pad, iodophor prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
  • Sterile Tyvek™ packaging
  • Come with Filter-Pro™ air bubble removal device
†With support kit.
Specifications
Needle Gauge 22 to 23 G
For Use With (Application) Provides consistent and accurate analysis for all parameters tested, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels
Includes Pulsator™ 3mL Luer lock syringe with Filter-Pro™ device, 22g x 1.5 in. and 23g x 1 in. needles, Needle-Pro device, support kit, needle stop, and polybag packaging
Product Type Heparin, 22, 23G Needle
Quantity 100/Cs.
Packaging Polybag
Volume (Metric) 3 mL