Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICU Medical Pulsator™ Plus Liquid Heparin Arterial Blood Sampling Kits

Click to view available options
Quantity:
100/Cs.
Product Type:
Heparin, 20, 22G Needle
Heparin, 22, 23G Needle
Packaging:
Polybag
Description
Includes
3-cc or 5-cc Luer lock syringe and Filter-Pro™ device in polybag or Tyvek packaging.
- Pulsating effect allows for precise positioning in arterial lumen
- Syringe response to arterial pressure helps ensure arterial sample
- Rapid fill minimizes patient discomfort
- 3mL arterial blood-sampling syringes are easily aspirated during low blood pressure or code situation
- Needles with different gauges and lengths
- Prefilled with liquid sodium heparin concentration of 31 IU/mL
- Specially formulated electrolyte-balanced liquid sodium heparin offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels
- Clot-free blood sampling without risking sample dilution
- Provides consistent and accurate analysis for all parameters tested
Order Info
- With or without support kit (alcohol prep pad, iodophor prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
- Sterile Tyvek™ packaging
- Come with Filter-Pro™ air bubble removal device
†With support kit.
Specifications
Specifications
Needle Gauge | 20 and 22 G |
For Use With (Application) | Provides consistent and accurate analysis for all parameters tested, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels |
Includes | Pulsator™ 3mL Luer lock syringe with Filter-Pro™ device, 20g x 1.5 in. and 22g x 1 in. needles, Needle-Pro™ device, support kit, needle stop, and polybag packaging |
Product Type | Heparin, 20, 22G Needle |
Quantity | 100/Cs. |
Packaging | Polybag |
Volume (Metric) | 3 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction