Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMMHC/Myosin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984343
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4343 Supplier Bioss Supplier No. BS1956R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen SMMHC/Myosin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYH11
Gene Accession No. P12882, P35749
Gene Alias AAT4; BOS_18904; BOS_22758; epididymis luminal protein 71; FAA4; HEL71; KIAA0866; MYH1; MYH11; MYHa; MyHC-2x; MyHC-2X/D; MyHC-IIx/d; MYHSA1; Myosin heavy chain 1; Myosin heavy chain 11; Myosin heavy chain 2x; Myosin heavy chain IIx/d; myosin heavy chain SMA; Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, smooth muscle isoform; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 11, smooth muscle; Myosin-1; Myosin-11; SMHC; SMMHC; type 2x myosin heavy chain
Gene Symbols MYH1, MYH11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SMHC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4619, 4629
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.