Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMN Antibody (2B1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1001936AF594
Description
SMN Monoclonal specifically detects SMN in Human, Mouse, Rat, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SMN | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
SMN1 | |
Purified recombinant His6-tagged human SMN protein. [Swiss-ProtNo. Q16637] | |
Primary | |
Human, Mouse, Rat, Primate, Xenopus species, Zebrafish | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2B1 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric | |
Mouse | |
0.1 mL | |
6606 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction