Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMN Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103326AF532
Description
SMN Polyclonal antibody specifically detects SMN in Human samples. It is validated for Western BlotSpecifications
SMN | |
Polyclonal | |
Western Blot | |
Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric | |
The immunogen recognized by this antibody maps to a region between residue 244 and 294 of human survival of motor neuron 1 using the numbering given in entry NP_000335.1 (GeneID 6606). | |
0.1 mL | |
Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators | |
6606 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction