Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMN Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SMN |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3831
![]() ![]() |
Proteintech Group Inc
223291AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The StARD (steroidogenic acute regulatory protein-related lipid transfer (START) domain containing) family of proteins is comprised of fifteen different members. All members contain the characteristic START domain and are believed to play key roles in the metabolism and transport of lipids. The StARD proteins are grouped into six subfamilies based on their START domain sequences. PC-TP (StARD2), StARD7, StARD10 and GPBP (StARD11) constitute one subfamily, namely the STARD2/PCTP group. StARD10, also known as PCTP2, PCTPL, NY-CO-28, CGI-52 or SDCCAG28, is widely expressed and functions in phospholipid transfer, binding to phosphatidylcholine and phosphatidylethanolamine. StARD10 can be found in sperm flagellum, potentially functioning as an enzyme involved in energy metabolism, and its expression is developmentally regulated in testis and mammary glands. StARD10 activity can be inhibited via phosphorylation by casein kinase II.Specifications
SMN | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q16637 | |
6606 | |
SMN Fusion Protein Ag17798 | |
Primary | |
-20°C | |
SMN1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C BCD541, Component of gems 1, Gemin 1, SMN, SMN1, SMN1,SMN, SMN2, SMNC, SMNT, Survival motor neuron protein | |
SMN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title