Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMNDC1 Antibody (PCRP-SMNDC1-1A9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308232B
Description
SMNDC1 Monoclonal antibody specifically detects SMNDC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
SMNDC1 | |
Monoclonal | |
Biotin | |
SMNRSMN-related protein, SPF30splicing factor 30, survival of motor neuron-related, survival motor neuron domain containing 1,30 kDa splicing factor SMNrp, Survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30 | |
Recombinant full-length human SMNDC1 protein (Uniprot: O75940 ) | |
100 μg | |
Apoptosis | |
10285 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-SMNDC1-1A9 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction